Key Documents
Z322202
Quickfit® right-angle tubing adapter
joint: ST/NS 29/32
Synonym(s):
MF10/4B/SC
Select a Size
1 EA
€36.70
€36.70
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€36.70
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€36.70
Please contact Customer Service for Availability
Joint
joint: ST/NS29/32
manufacturer/tradename
Quickfit MF 10/4B/SC
availability
not available in USA and Canada
not available in USA and Canada
General description
With joint and right angle-connection with screw-thread connector for flexible tubing.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)