Key Documents
Z314994
Hirschmann® pipetus®-Standard pipette controller
with pump and sterile filter
Select a Size
1 EA
€344.00
1 EA
€344.00
1 EA
€410.00
€344.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€344.00
1 EA
€344.00
1 EA
€410.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€344.00
Please contact Customer Service for Availability
AC/DC input
230 V AC
material
plastic
silicone
feature
CE compliant
manufacturer/tradename
Hirschmann 9904000
availability
not available in USA and Canada
volume
0.1-200 mL
General description
Similar to the Pipetus-Akku, but with separate pump unit. The separate pumps avoids any contamination and lowers ignition risk. There are no electronic parts within the controller.
Supplied with tubing, hand-held controller, pipette holder, spare filters, spare safety valve and instruction manual.
UK & European version supplied with 2-pin plug and 3-pin adapter.
Sterile filter mounted on pump, provides clean sterile airflow if required.
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Pipetus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)