Key Documents
Z309400
Quickfit® flat-bottom boiling flasks
capacity 250 mL, joint: ST/NS 29/32
Synonym(s):
FF250/4S
Select a Size
1 EA
€41.90
5 EA
€156.00
€41.90
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€41.90
5 EA
€156.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€41.90
Please contact Customer Service for Availability
material
borosilicate glass
flat bottom flask
short
single-neck
Joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FF 250/4S
availability
not available in USA and Canada
capacity
250 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)