Z309257
Quickfit® three-neck round-bottom flasks with angled necks
capacity 100 mL, center joint: ST/NS 24/29, side joint: ST/NS 14/23
Synonym(s):
FR100/3S/11A
Select a Size
€72.20
Select a Size
About This Item
€72.20
Recommended Products
material
borosilicate glass
round bottom flask
three angled neck
Joint
center joint: ST/NS24/29
side joint: ST/NS14/23
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Quickfit FR100/3S/11A
availability
not available in USA and Canada
H
110 mm
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
- One vertical neck
- Two angled side necks
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service