Key Documents
Z303925
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 19/26, parallel socket joint: 14/32, 5° side socket joint: 14/23, 10° side socket joint: 14/23, 15° side socket joint: 29/32
Synonym(s):
MAF2/41
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass
label
C
Joint
10° side socket joint: 14/23
15° side socket joint: 29/32
5° side socket joint: 14/23
center joint: ST/NS19/26
parallel socket joint: 14/32
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Quickfit MAF 2/41
availability
not available in USA and Canada
General description
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)