Z303518
Quickfit® three-neck round-bottom flasks with angled necks
capacity 100 mL, center joint: ST/NS 24/29, side joint: ST/NS 19/26
Synonym(s):
FR100/3S/22A
About This Item
Recommended Products
material
borosilicate glass
round bottom flask
three angled neck
Joint
center joint: ST/NS24/29
side joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Quickfit FR100/3S/22A
availability
not available in USA and Canada
H
110 mm
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
- One vertical neck
- Two angled side necks
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service