Skip to Content
Merck
All Photos(1)

Key Documents

Z302430

Quickfit® Cone/Screw-thread Adapters

24 thread (couples 10.5 - 11.5 mm diameter), joint: ST/NS 19/26

Synonym(s):

ST52/24

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€40.60

€40.60


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€40.60

About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

€40.60


Please contact Customer Service for Availability

Joint

joint: ST/NS19/26

manufacturer/tradename

Quickfit ST 52/24

availability

not available in USA and Canada
not available in USA and Canada

fitting

24 thread (couples 10.5 - 11.5 mm diameter)

Looking for similar products? Visit Product Comparison Guide

General description

The Quickfit screw-thread joints on these cone adapters provide a versatile means of coupling thermometers, bleed tubes, electrodes, etc; with Quickfit glassware. The joints are readily adjustable for depth of immersion by slackening the screw cap, and give excellent results under vacuum.

Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.

For replacement parts, see related products section of web site listing.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service