Key Documents
Z301116
Quickfit® expansion adapter
top female joint: ST/NS 24/29, bottom male joint: ST/NS 19/26
Synonym(s):
XA32
Select a Size
1 EA
€33.00
€33.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€33.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€33.00
Please contact Customer Service for Availability
Joint
female joint: ST/NS top24/29
male joint: ST/NS bottom19/26
manufacturer/tradename
Quickfit XA 32
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)