Skip to Content
Merck
All Photos(1)

Key Documents

Z284688

Supelco

Spectroline® battery-operated UV lamp

Replacement 4 W longwave bulb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

technique(s)

thin layer chromatography (TLC): suitable

size

6 1/4 in. × 2 in. × 1 in.

Looking for similar products? Visit Product Comparison Guide

General description

  • 4 W longwave bulb
  • Requires 4 AA batteries (not included)

Footnote

CE Approved

Legal Information

Spectroline is a registered trademark of Spectronics, Corp.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service