Documents
Z283835
Spinbar® magnetic stirring fleas
white, diam. 3 mm, L 3 mm
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
material
PTFE-coated
cylindrical PTFE-coated
feature
ALNICO V coated magnet
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F37119-0003
L
3 mm
diam.
3 mm
color
white
General description
  • Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
  • Ideal for mixing of small volume samples in environmental testing and life science applications.
  • Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
  • Broad chemical resistance for use in a wide variety of applications.
  • Autoclavable.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company