Key Documents
Z280410
Sigma-Aldrich
PVC laboratory tubing
I.D. × O.D. 1/2 in. × 5/8 in.
Synonym(s):
Saint Gobain Versilon ABH02036
Select a Size
100 FT
€86.10
€86.10
Please contact Customer Service for Availability
Select a Size
Change View
100 FT
€86.10
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
€86.10
Please contact Customer Service for Availability
packaging
pkg of 100 feet
I.D. × O.D.
1/2 in. × 5/8 in.
wall size
1/16 in.
General description
General-purpose polyvinyl chloride laboratory tubing. Clear, flexible tubing ideal for drains and rinse tubes. Max. temp.: 165°F (73.4°C).
Choose from one of the most recent versions:
Certificates of Analysis (COA)