Key Documents
Z277827
Sigma-Aldrich
HGS Space-filling molecular models
Researcher set
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
manufacturer/tradename
HGS 2001
General description
Designed for building biomolecules and complexed molecules such as steroids and peptides. The idea of α-helix or β -sheet in peptide molecules can be visualized. Suited for studying biosynthesis through nucleotides. 383 pieces.
These models combine the idea of both ball-and-stick and space-filling models for visualization of molecular frameworks as well as molecular structures covered with electron clouds. Color-coded model parts are made of flexible polyacetal covered with transparent polyacrylate. Can be used interchangeably with HGS biochemistry molecular models.
Elementary set= 137 pieces
Research set= 383 pieces
Scale: 1 Å = 1.0 cm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)