Skip to Content
Merck
All Photos(1)

Key Documents

Z273244

Sigma-Aldrich

Alcotabs® efferescent tablets for siphon-type pipette and test tube washers

For siphon-type pipette and test-tube washers

Synonym(s):

efferescent detergent tablet, tube washer detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€52.10
6 X 100 TABS
€250.00

€52.10


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€52.10
6 X 100 TABS
€250.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

€52.10


Please contact Customer Service for Availability

packaging

pkg of 6 x 100 tablets in 6PAK
pkg of 100 tablets in 1PAK

manufacturer/tradename

Alconox, Inc. 1500 (6 x 100 Tablets)
Alconox, Inc. 1500-1 (100 Tablets)

General description

For siphon-type pipette and test-tube washers.

Legal Information

Alcotabs is a registered trademark of Alconox, Inc.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service