Key Documents
Z266280
Sigma-Aldrich
Polymer-body combination pH electrodes
BNC fitting
Select a Size
1 EA
€333.00
€333.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€333.00
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
€333.00
Please contact Customer Service for Availability
stem L × diam.
183 mm × 6 mm
General description
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Sealed, gel-filled
Long, narrow/tubes, vials.
The combination of a pH indicating and reference half-cell in an impact-resistant epoxy body. Designed for applications where breakage is a problem.
  • Environmental - field sampling
  • Industrial - process control
  • Educational - core courses
Rugged bulb, fast response in optimal range of 0 to 12 pH. Suggested temp. range: 0 to 80 °C. For Tris, sulfides, and heavy metals, choose a double junction reference electrode. Supplied with 36 in. lead.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)