Key Documents
Z256331
Wrap-it ties
L 7 1/2 in.
Synonym(s):
tie wrap, wrap tie
Select a Size
250 EA
€54.70
€54.70
Please contact Customer Service for Availability
Select a Size
Change View
250 EA
€54.70
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
€54.70
Please contact Customer Service for Availability
parameter
50 lb tensile strength
L
7 1/2 in.
General description
Self-locking, one-piece nylon construction. Excellent for securing rubber septa to bottles and flask joints, attachment of tubing to hose barbs or glassware, and for closing plastic bags or drum liners. Use with Wrap-it tie installing tool.
Tear-off version is blue in color and is easily removed from septum with a quick pulling motion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)