Skip to Content
Merck
All Photos(1)

Key Documents

Z252077

Sigma-Aldrich

Nalgene® polypropylene screw-closure for dropper-dispenser bottles

closure size 24 mm

Synonym(s):

bottle closure, plastic screw cap

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€10.50

€10.50


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€10.50

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

€10.50


Please contact Customer Service for Availability

packaging

pkg of 12 ea

manufacturer/tradename

Nalgene 712150-0240

closure size

24 mm

Looking for similar products? Visit Product Comparison Guide

General description

Replacement polypropylene closures for use with Nalgene® narrow-mouth bottles, 24mm.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service