Key Documents
Z244112
Sigma-Aldrich
Sample cylinder plug
brass
Select a Size
5 EA
€106.00
€106.00
Please contact Customer Service for Availability
Select a Size
Change View
5 EA
€106.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
€106.00
Please contact Customer Service for Availability
material
brass
General description
The 1/4 in. NPTM square head plug fits all Sure/Pac sample cylinder sizes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)