Z231665
Duran® crystallizing dishes
capacity 20 mL
Synonym(s):
213132403
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
packaging
pack of 1 ea
case of 10 ea
manufacturer/tradename
DURAN® 213132403
availability
not available in USA and Canada
capacity
20 mL
dish diam.
40 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Cylindrical dishes without spout.
- Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
- The design of the dishes allow solvents to evaporate quickly and evenly
- Fired-on, highly durable white ceramic
- Meets DIN 12337 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service