Key Documents
Z222356
Sigma-Aldrich
Keck® Joint Clip
standard taper, joint size 45, brown
Select a Size
12 EA
€125.00
€125.00
Please contact Customer Service for Availability
Select a Size
Change View
12 EA
€125.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€125.00
Please contact Customer Service for Availability
color
brown
General description
Color coded, Delrin® plastic joint clips will not scratch glassware or corrode. Withstand temperatures up to 302 °F (150 °C). Resistant to concentrated alkalis and diluted acids; not resistant to halogen gases or concentrated acids.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Keck is a registered trademark of Adcuram Lab Technologies GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)