Key Documents
Z219916
Sigma-Aldrich
Aldrich® hollow pennyhead stopper
joint: ST/NS 10/30
Synonym(s):
glass stopper, glass stoppers, stoppers
Select a Size
1 EA
€25.20
€25.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€25.20
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
€25.20
Please contact Customer Service for Availability
material
ground glass
Joint
joint: ST/NS10/30
description
closed at bottom
General description
Conforms to standard-taper specifications as outlined in ASTM E 675.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)