Key Documents
Z219827
Sigma-Aldrich
Single-bulb ball-tube
capacity 1,000 mL, joint: ST/NS 24/40, w/drip-tip
Select a Size
1 EA
€204.00
€204.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€204.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
€204.00
Please contact Customer Service for Availability
Joint
joint: ST/NS24/40
capacity
1,000 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)