Key Documents
Z203246
Sigma-Aldrich
Aldrich® anticlimb splash-guard adapter
female joint: ST/NS 29/32, male joint: ST/NS 29/32, capacity 250 mL
Select a Size
1 EA
€197.00
€197.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€197.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€197.00
Please contact Customer Service for Availability
Joint
female joint: ST/NS29/32
male joint: ST/NS29/32
capacity
250 mL
Related Categories
General description
Aldrich® anticlimb splash-guard adapter greatly lower carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser.
Application
Aldrich® anticlimb splash-guard adapter are ideal for use with solvents that foam excessively during rotary evaporation. Inner vapor tube is sealed to the top of trap to prevent foam entry into condenser.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)