Key Documents
Z164011
Sigma-Aldrich
Aldrich® fractional distillation head
with Vigreux indents
Select a Size
1 EA
€1,080.00
€1,080.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€1,080.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
€1,080.00
Please contact Customer Service for Availability
Related Categories
General description
<ST/>14/20 and 24/40 male joints and <ST/>14/20 female joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)