Key Documents
Z154075
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 35/20
Select a Size
1 EA
€49.70
€49.70
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€49.70
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€49.70
Please contact Customer Service for Availability
material
stainless steel
Joint
joint: SJ35/20
General description
For use with Z137294, and for Ace pressure filter/drying funnels.
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)