Key Documents
Z146544
Sigma-Aldrich
Ace Rotary-Evaporator/Freeze-Drying Flask Adapter Head W/ O-ring
joint: ST/NS 24/40
Select a Size
1 EA
€258.00
€258.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€258.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€258.00
Please contact Customer Service for Availability
Joint
joint: ST/NS24/40
description
with FETFE®
O-ring.
manufacturer/tradename
Ace Glass 703021
General description
Flange with two ground flats opposite each other allows nylon retainers on adapter head to pass over. O-ring in head and flask grooves make a vacuum-tight seal when head is turned 90° in either direction. No grease, springs, or clamp necessary. Flasks sold separately.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)