Key Documents
Z146218
Supelco
Aldrich® rectangular TLC developing tanks, lid only
For Z126195
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
technique(s)
thin layer chromatography (TLC): suitable
General description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results.[1] Lid is used to cover the tank.
Application
Aldrich® rectangular TLC developing tanks, lid only may be used to develop TLC plates.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
An overview on thin layer chromatography
Bele, Archana A., and Anubha Khale.
International Journal of Pharmaceutical Sciences and Research, 2, 256-267 (2011)