Z131377
Nalgene® filtering flask
capacity 500 mL, stopper joint: 7
Synonym(s):
polypropylene vacuum flask, vacuum flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
flat bottom flask
polypropylene
Joint
joint ST/NS neckstopper #7
joint: stopper7
sterility
non-sterile
feature
closure type Stopper
packaging
pack of 1
manufacturer/tradename
Nalgene® DS4101-0500
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Nalgene® filtering flasks are an excellent alternative to glass flasks.
- Manufactured from light-weight, autoclavable, polypropylene that is resistant to shattering.
- Molded-in one-piece to withstand 1 atm vacuum.
- Tubulation at a 45° angle helps prevent tipping.
- Barbed adapter accepts 1/4 to 3/8 in. I.D. tubing. (sold separately)
- Oval marking area facilitates easy labeling of contents.
- Neck sized to fit standard stopper sizes (stopper not included).
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service