Key Documents
Z125342
Ace PTFE ferrules
for tubing o.d., 12.7 mm (1/2 in.)
Select a Size
12 EA
€60.30
€60.30
Please contact Customer Service for Availability
Select a Size
Change View
12 EA
€60.30
About This Item
UNSPSC Code:
27121703
NACRES:
NB.35
€60.30
Please contact Customer Service for Availability
feature
for tubing o.d. 12.7 mm (1/2 in.)
manufacturer/tradename
Ace Glass 1171015
Related Categories
General description
For use in place of O-rings on threaded glassware.
Choose from one of the most recent versions:
Certificates of Analysis (COA)