Key Documents
Z121525
Sigma-Aldrich
Aldrich® cold-trap with ball joint
joint: SJ 75/50
Select a Size
1 EA
€854.00
€854.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€854.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
€854.00
Please contact Customer Service for Availability
Joint
joint: SJ75/50
Related Categories
General description
Trap 7 cm I.D. × 30 cm Length, overall 75 mm O.D. × 470 mm Length, 20 mm O.D. side tubes. Includes pinch clamp.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)