Z118923
Safety bulb®
pkg of 1 ea
Synonym(s):
pipette bulb, pipette filler
All Photos(1)
About This Item
Recommended Products
material
rubber
silicone
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H37888-0000
Related Categories
General description
Large capacity, for all pipette sizes. Tapered silicone seal prevents loss of suction. May be opened for cleaning; autoclavable. Elastic cord provided for hanging on reagent bottles.
Legal Information
Safety bulb is a registered trademark of SP Scienceware, a division of SP