Key Documents
Z106534
Supelco
Septa for open top caps
for cap size 15 mm, PTFE/red rubber
Select a Size
100 EA
€127.00
€127.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
100 EA
€127.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
€127.00
Please contact Customer Service for Availability
Request a Bulk Order
material
PTFE/red rubber
packaging
pkg of 100
General description
For use with cut-out top caps and Wheaton vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)