WHA67205001
Whatman® Polydisc TF Chemical resistant in-line filter
pore size 0.1 μm
Synonym(s):
Whatman filter, Z744247, in line filter, line filter
Select a Size
€170.00
Select a Size
About This Item
€170.00
Recommended Products
material
PTFE membrane
polypropylene housing
sterility
non-sterile
packaging
pkg of 10 ea
manufacturer/tradename
Whatman 6720-5001
Whatman Article No. 28420211 (US reference)
diam.
50 mm
pore size
0.1 μm
Looking for similar products? Visit Product Comparison Guide
General description
Polydisc TF aggressive solvent filter
Filtering organic solvents and aggressive solutions calls for a chemically resistant, durable membrane. Cytiva′s Polydisc TF features a PTFE membrane resistant to such compounds. The 1 µm membrane features a polypropylene prefilter for solvent filtration containing high particulate density in order to extend the useful life of the membrane.
Application
- Pharmaceutical: vents and in-line applications
- Biotech: sterile vents and exhausts for growth environments, in-line sterilization of gases
- Laboratory: clean or sterile gases. Filtration of solvents and reagents. Drying gases.
- Electronics: photoresists, solvents, gases for research
Features and Benefits
- Chemically resistant hydrophobic PTFE membrane is well suited as a solvent filter for many different solutions
- Filter can be autoclaved multiple times without losing integrity
- Integrity-testable (bubble point or water breakthrough pressure in situ methods)
- Lightweight to prevent tube collapsing
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service