Skip to Content
Merck
All Photos(1)

Key Documents

WHA30306132

Whatman® cellulose chromatography papers

3MM CHR, W × L 12 cm × 14 cm, pkg of 100 ea

Synonym(s):

Whatman paper, Z742325, blotting papers, cellulose membranes

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€31.50

€31.50


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€31.50

About This Item

UNSPSC Code:
41115727
NACRES:
NB.24

€31.50


Please contact Customer Service for Availability

material

cellulose membrane
sheets

sterility

non-sterile

feature

3MM CHR

packaging

pkg of 100 ea

manufacturer/tradename

Whatman 3030-6132
Whatman Article No. 28419265 (US reference)

parameter

130 mm/30 min flow rate

W × L

12 cm × 14 cm

basis weight

185 g/m2

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

General description

0.34 mm Whatman medium thickness 3MM Chr paper. Whatman 3MM Chr paper is the world’s most widely used blotting paper. This acceptance and usage reflect the high quality, purity, and consistency that are relied upon by researchers doing Southern, Northern, and Western transfers. 3MM Chr paper is now available in the most widely used sizes. A medium thickness paper (0.34 mm) is used extensively in electrophoresis for lifting of sequencing gels. 3MM Chr is also used both in electrophoresis and for general chemistry. A medium thickness paper (0.34 mm) used extensively for general chromatography and electrophoresis.

Features and Benefits

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind. Ensures that no contamination will occur during the transfer steps.
  • Manufactured and tested specifically for chromatographic and blotting techniques. This ensures the wicking capability and uniformity of capillary action that are important in obtaining clean and even transfers during blotting.
  • Whatman 3MM Chr is considered the industry standard for blotting procedures.
  • Convenient sizes available in sheets precisely cut to the most popular gel and transfer membrane sizes. Allows "out-of-the-box" usage and eliminates sheet-to-sheet variations.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service