WHA2105862
Whatman® lens cleaning tissue, Grade 105
size 200 mm × 300 mm, pkg of 100 ea
Synonym(s):
Z741361, lens cleaning paper, lens paper, lens tissue
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 2105-862
Whatman Article No. 28418696 (US reference)
size
200 mm × 300 mm
General description
High-quality whatman lens cleaning tissue, which is chemically pure and free from silicones and other additives. Lenses and other optical surfaces made from glass, quartz or plastic can be easily scratched if you do not clean them with a very soft surface. High quality Whatman lens cleaning tissue provides the solution. The tissue is chemically pure and free from silicones and other additives. Most importantly, it can be relied on to safely remove surface moisture and grease.
Features and Benefits
- Soft texture will not damage lenses or optical surfaces.
- Chemically pure tissue is free from silicones and other additives.
- High absorbency leads to increased safety upon removal of surface moisture and grease.
- Very strong and leaves no fibers.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva