WHA2017006
Whatman® Antibiotic Assay Discs
diam. 6 mm
Synonym(s):
Whatman paper, Z741310, microbial assays
Select a Size
€186.00
Select a Size
About This Item
€186.00
Recommended Products
packaging
pkg of 1000 ea
manufacturer/tradename
Whatman 2017-006
Whatman Article No. 28418660 (US reference)
diam.
6 mm
General description
Features and Benefits
- The antibiogram allows rational and selective chemotherapy.
- Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
- The size of the inhibition zone is a measure for the effectiveness of the substances.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)