Documents
WHA10483064
Whatman® hybridization reaction folders
Synonym(s):
Z698962
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 10483064
Whatman Article No. 28416120 (US reference)
General description
Sealable hybridization bags
Whatman reaction folders are sealable bags for hybridization and incubation reactions.
The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags. The folders can be sealed with a standard heat-sealer. Available in 8 x 9.5 inch (20.32 x 24.13 cm) size.
Features and Benefits
  • The folders are open on three sides to allow easier insertion of a wet membrane when compared to standard hybridization bags
  • The folders can be sealed with a standard heat-sealer.
  • Available in 203.2 x 241.3 mm (8 x 9.5 in) size.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva