WHA10442002
Whatman® glass vacuum filtration device
GV050/0/02, lower part for GV050/0, 1/pk
Synonym(s):
Z698318, glass vacuum filtration unit, vacuum filtration system
Select a Size
€101.00
Select a Size
About This Item
€101.00
Recommended Products
material
borosilicate glass funnel(s) (funnel)
borosilicate glass housing
stainless steel clamp(s) (clamp)
feature
lower part for GV050/0
packaging
pack of 1
manufacturer/tradename
Whatman 10442002
Whatman Article No. 28415553 (US reference)
suitability
suitable for (filtration and biological analysis; the 304 stainless steel support screen is suitable for use with proteinaceous solutions)
Related Categories
General description
Produced from borosilicate glass and available with a choice of support screen. Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
The sintered glass support is recommended for filtration and biological analysis. The 304 stainless steel support screen is suitable for use with proteinaceous solutions.
Features and Benefits
- Accessory to GV050 Glass Filter Funnel
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)