WHA10410014
Whatman® regenerated cellulose RC60 filter discs 1 μm pore size
white, hydrophilic, 100 ea, 50 mm diam
Synonym(s):
Z697885, filter membrane, membrane filter, syringe filter
Select a Size
€334.00
Select a Size
About This Item
€334.00
Recommended Products
Product Name
Whatman® regenerated cellulose membrane filters, RC60 Membrane Circles, diam. 50 mm, pore size 1 μm
feature
hydrophilic
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 10410014
Whatman Article No. 28415270 (US reference)
diam.
50 mm
pore size
1 μm
Related Categories
General description
Versatile regenerated cellulose syringe filter membranes
Syringe filters require high-quality microporous membrane filters that minimally impact sample analysis with extractable impurities. Regenerated cellulose membrane filters from Cytiva′s business have broad solvent compatibility, and they contribute very low extractable material in a wide variety of sample solvents. Thus, they are appropriate for sample preparation in many applications and as a standalone or syringe filter membrane.
Features and Benefits
- Spontaneous wetting with very good wet strength
- Chemical resistance suitable for a wide range of aqueous and organic media
- Mechanically stable with low protein binding
- Low extractable levels to minimize sample contamination
- Can be sterilized by all common methods
Other Notes
Legal Information
Sold under license to DE10102744 and foreign equivalents thereof
Choose from one of the most recent versions:
Certificates of Analysis (COA)