WHA10404090
Whatman® Elutrap electroelution system
ELUTRAP BT-1 Membranes
Synonym(s):
Z697478, electro elution, elutrap
All Photos(1)
About This Item
UNSPSC Code:
41104900
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 10404090
Whatman Article No. 28415140 (US reference)
General description
The Elutrap System is designed to isolate nucleic acids and proteins from agarose or polyacrylamide gel slices by electroelution. Samples are purified with excellent recovery into volumes as low as 200 μL, without requiring sample pretreatment or special buffers.
The Elutrap System can be used with most horizontal gel electrophoresis chambers. The Elutrap Electrophoresis Chamber allows for the most efficient flow of current through the device and can be used for up to four samples simultaneously.
Assembly of the Elutrap System is very easy. Gel slices are placed in the middle of the Elutrap device, which is then placed into a horizontal electrophoresis chamber. Molecules migrate from the gel slice into a trap area formed by BT1 and BT2 membranes. The membrane placement is adjustable, allowing final elution trap volumes to be optimized for the particular assay. The Elutrap System can also be used for concentration of dilute solutions.
Features and Benefits
The Elutrap System can be used with most horizontal gel electrophoresis chambers. The Elutrap Electrophoresis Chamber allows for the most efficient flow of current through the device and can be used for up to four samples simultaneously.
Assembly of the Elutrap System is very easy. Gel slices are placed in the middle of the Elutrap device, which is then placed into a horizontal electrophoresis chamber. Molecules migrate from the gel slice into a trap area formed by BT1 and BT2 membranes. The membrane placement is adjustable, allowing final elution trap volumes to be optimized for the particular assay. The Elutrap System can also be used for concentration of dilute solutions.
Features and Benefits
- Versatile system can be used for nucleic acids and proteins
- Purifies nucleic acids 14 bp to 150 Kb; proteins larger than 3 to 5 kD
- No salt cushions or special buffers required for elution
- Adjustable trap allows optimization of final sample volume
- Electrophoresis chamber holds up to four Elutrap devices simultaneously
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)