WHA10310245
Whatman® application specific filter, prepleated qualitative filter paper, Grade 287 1/2
circles, diam. 150 mm, pack of 50
Synonym(s):
Z749044
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cellulose membrane (Kieselguhr paper)
circles
sterility
non-sterile
feature
Grade 287 1/2
packaging
pack of 50
manufacturer/tradename
Whatman 10310245
Whatman Article No. 28414165 (US reference)
parameter
330 sec/100 mL speed (Herzberg)
diam.
150 mm
thickness
360 μm
basis weight
161 g/m2
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles:
Grade 287 ½: kieslghur paper
Application specific cellulose filter paper. Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
- Increased total loading capacity as more filter area is available
- Maintained flow rate due to the reduction in filter paper contact with funnel side and, the self-supporting shape of the filter itself
Grade 287 ½: kieslghur paper
Application specific cellulose filter paper. Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.
Features and Benefits
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
- Increased total loading capacity as more filter area is available.
- Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
- The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service