Skip to Content
Merck
All Photos(1)

Key Documents

W6837

Fumaric acid-1,4-13C2

API for Clinical Studies, ≥99 atom % 13C, ≥98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOO13CCH=CH13COOH
Molecular Weight:
118.06
MDL number:
UNSPSC Code:
12352115

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

mp

298-300 (subl) (lit.)

mass shift

M+2

storage temp.

15-25°C

SMILES string

O[13C](/C=C/[13C](O)=O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+/i3+1,4+1

InChI key

VZCYOOQTPOCHFL-ZWWDXPBASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

446.0 °F - closed cup

Flash Point(C)

230 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service