Skip to Content
Merck
All Photos(1)

Key Documents

W508209

Sigma-Aldrich

3-Methyl-3-pentanol

≥98%, FG

Synonym(s):

Diethyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2C(CH3)OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731456
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.184
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.418 (lit.)

bp

123 °C (lit.)

mp

−38 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity

SMILES string

CCC(C)(O)CC

InChI

1S/C6H14O/c1-4-6(3,7)5-2/h7H,4-5H2,1-3H3

InChI key

FRDAATYAJDYRNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-3-pentanol is an aroma-active alcohol that occurs naturally in pandan leaves, red pepper and fruit of Lycii fructus.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aroma?Active Components of Lycii fructus (kukija).
Lee GH, et al.
Journal of Food Science, C500?C505-C500?C505 (2008)
Volatile composition of pandan leaves (Pandanus amaryllifolius).
Jiang J.
Flavor chemistry of ethnic foods, 105-109 (1999)
Volatiles from red pepper (Capsicum spp.).
Keller U, et al.
ACS Symp. Ser., 137?146-137?146 (1981)
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service