Skip to Content
Merck
All Photos(1)

Key Documents

W481911

Sigma-Aldrich

meso-Erythritol

≥99%, FG

Synonym(s):

1,2,3,4-Butanetetrol, meso-1,2,3,4-Tetrahydroxybutane, i-Erythritol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
FEMA Number:
4819
Beilstein:
1719753
MDL number:
UNSPSC Code:
41116107
E Number:
E 968
NACRES:
NA.21
Organoleptic:
odorless
grade:
FG
Halal
Kosher
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

bp

329-331 °C (lit.)

mp

118-120 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC[C@@H](O)[C@@H](O)CO

InChI

1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+

InChI key

UNXHWFMMPAWVPI-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Allelic variation of the Tas1r3 gene affects behavioral taste responses to this sugar alcohol, suggesting that it is a T1R3 receptor ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service