Skip to Content
Merck
All Photos(1)

Key Documents

W423501

Sigma-Aldrich

ε-Caprolactam

Synonym(s):

2-Oxohexamethyleneimine, Aza-2-cycloheptanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
FEMA Number:
4235
Beilstein:
106934
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.052

biological source

synthetic

vapor pressure

<0.01 mmHg ( 20 °C)

autoignition temp.

707 °F

expl. lim.

8 %

bp

136-138 °C/10 mmHg (lit.)

mp

68-71 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

O=C1CCCCCN1

InChI

1S/C6H11NO/c8-6-4-2-1-3-5-7-6/h1-5H2,(H,7,8)

InChI key

JBKVHLHDHHXQEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgeta Voicu et al.
International journal of pharmaceutics, 446(1-2), 63-69 (2013-02-14)
Here, we report the fabrication of a novel ε-caprolactam-silica (ε-SiO2) network and assessed its biocompatibility and ability to improve the antimicrobial activity of kanamycin. The results of the quantitative antimicrobial assay demonstrate that the obtained ε-SiO2 network has efficiently improved
Andrew J Niehaus et al.
American journal of veterinary research, 74(3), 381-385 (2013-02-27)
To compare the mechanical characteristics of polymerized caprolactam and monofilament nylon loops with those of the cranial cruciate ligament (CCL) in cattle. 6 femorotibial joints harvested from 3 cows and suture constructs made from No. 8 polymerized caprolactam, 80-lb test
Ahmad Bitar et al.
Journal of biomedical nanotechnology, 8(5), 709-719 (2012-08-15)
Temperature and glucose sensitive microgels were prepared via radical precipitation polymerization, using N-vinylcaprolactam (NVCL) as the main monomer, N,N-methylenebisacrylamide (MBA) as a crosslinker, V50 as initiator and 4-vinylphenylboronic acid (VPBA) as a functional monomer. The LCST of homopoly (NVCL) was
Juliana S Félix et al.
Analytical and bioanalytical chemistry, 403(10), 2869-2882 (2012-04-25)
Adhesives used in food packaging to glue different materials can provide several substances as potential migrants, and the identification of potential migrants and migration tests are required to assess safety in the use of adhesives. Solid-phase microextraction in headspace mode
Alla N Generalova et al.
Biosensors & bioelectronics, 39(1), 187-193 (2012-08-14)
Composite polymer particles consisting of a solid poly(acrolein-co-styrene) core and a poly(N-vinylcaprolactam) (PVCL) polymer shell doped with CdSe/ZnS semiconductor quantum dots (QDs) were fabricated. The temperature response of the composite particles was observed as a decrease in their hydrodynamic diameter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service