Skip to Content
Merck
All Photos(1)

Key Documents

W419301

Sigma-Aldrich

trans-2-Pentenoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCO2H
CAS Number:
Molecular Weight:
100.12
FEMA Number:
4193
Beilstein:
1720312
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.107
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

98%

impurities

≤10% cis isomer

refractive index

n20/D 1.452 (lit.)

bp

106 °C/20 mmHg (lit.)

mp

9-11 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; cheesy

SMILES string

[H]\C(CC)=C(\[H])C(O)=O

InChI

1S/C5H8O2/c1-2-3-4-5(6)7/h3-4H,2H2,1H3,(H,6,7)/b4-3+

InChI key

YIYBQIKDCADOSF-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayana Dagan-Wiener et al.
Scientific reports, 7(1), 12074-12074 (2017-09-25)
Bitter taste is an innately aversive taste modality that is considered to protect animals from consuming toxic compounds. Yet, bitterness is not always noxious and some bitter compounds have beneficial effects on health. Hundreds of bitter compounds were reported (and
Philipp Höfer et al.
Microbial cell factories, 9, 70-70 (2010-09-18)
Methylotrophic (methanol-utilizing) bacteria offer great potential as cell factories in the production of numerous products from biomass-derived methanol. Bio-methanol is essentially a non-food substrate, an advantage over sugar-utilizing cell factories. Low-value products as well as fine chemicals and advanced materials
Ayse Guventurk et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 81(1), 21-28 (2020-04-16)
Food industry wastewater (FIWW) streams with high organic content are among the most suitable and inexpensive candidates for polyhydroxyalkanoate (PHA) biopolymer production. Due to its high organic acid content, pickle industry wastewater (PIWW), can be considered as one of the
Ruifeng Li et al.
Nature chemical biology, 14(7), 664-670 (2018-05-23)
Introduction of innovative biocatalytic processes offers great promise for applications in green chemistry. However, owing to limited catalytic performance, the enzymes harvested from nature's biodiversity often need to be improved for their desired functions by time-consuming iterative rounds of laboratory
Brian D Hudson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(12), 4951-4965 (2012-08-25)
When it is difficult to develop selective ligands within a family of related G-protein-coupled receptors (GPCRs), chemically engineered receptors activated solely by synthetic ligands (RASSLs) are useful alternatives for probing receptor function. In the present work, we explored whether a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service