Skip to Content
Merck
All Photos(1)

Key Documents

W394300

Sigma-Aldrich

o-Anisic acid

≥99%

Synonym(s):

2-Methoxybenzoic acid, O-Methylsalicylic acid, o-Anisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3943
Beilstein:
509929
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Quality Level

Assay

≥99%

mp

98-100 °C (lit.)

application(s)

flavors and fragrances

SMILES string

COc1ccccc1C(O)=O

InChI

1S/C8H8O3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ILUJQPXNXACGAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

291.2 °F - closed cup

Flash Point(C)

144 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Böttcher et al.
Analytical chemistry, 79(4), 1507-1513 (2007-02-15)
The coupling of liquid chromatography to electrospray ionization quadrupole time-of-flight mass spectrometry can be a powerful tool for metabolomics, i.e., the comprehensive detection of low molecular weight compounds in biological systems. There have, however, been doubts about the feasibility and
M Gil et al.
Biodegradation, 11(1), 49-53 (2001-02-24)
Aromatic carboxylic acids substituted with methoxylated groups are among the most abundant products in "alpechin", the wastes resulting from pressing olives to obtain olive oil. Degradation of o-methoxybenzoate by an stable consortium made of a gram positive bacterium, Arthrobacter oxydans
T Sasaki et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 50(5), 905-909 (1999-04-24)
For in vivo measurement of the hydroxyl radical (.OH), we synthesized [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid by carboxylation of 2-bromomagnesiumanisol using [11C]CO2. The radiochemical yield of [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid calculated from trapped [11C]CO2 in
Quan Zhou et al.
The Journal of organic chemistry, 73(20), 8049-8056 (2008-09-20)
Mander reductive alkylation of methyl 2-methoxybenzoate with prenyl bromide and hydrolysis of the enol ether afforded methyl 6-oxo-1-prenyl-2-cyclohexenecarboxylate. This was converted in five steps (reduction of the ketone, saponification, iodolactonization, ozonolysis, and intramolecular aldol reaction) to a spiro lactone cyclopentenal.
Chun-Juan Dong et al.
Planta, 240(4), 687-700 (2014-07-19)
Salicylic acid (SA) is an important plant hormone, and its exogenous application can induce tolerance to multiple environmental stresses in plants. In this study, we examine the potential involvement of endogenous SA in response to chilling in cucumber (Cucumis sativus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service