Skip to Content
Merck
All Photos(1)

Key Documents

W391808

Sigma-Aldrich

4-tert-Butylphenol

≥99%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OH
CAS Number:
Molecular Weight:
150.22
FEMA Number:
3918
Beilstein:
1817334
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.064

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012

vapor pressure

1 mmHg ( 70 °C)

Assay

≥99%

bp

236-238 °C (lit.)

mp

96-101 °C (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless

density

0.908 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

leather

SMILES string

CC(C)(C)c1ccc(O)cc1

InChI

1S/C10H14O/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7,11H,1-3H3

InChI key

QHPQWRBYOIRBIT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylphenol is an alkylphenolic compound that can be used as a flavoring agent. It is also a component of adhesives and polymers used for food packaging.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Lei et al.
Ecotoxicology (London, England), 19(7), 1268-1276 (2010-06-17)
Landfill leachates contain a large amount of unknown harmful compounds derived from domestic and industrial sources. A toxicity effect-directed approach was used to identify biologically active compounds in three landfill leachate samples (S1-S3) by combining the Microtox test with reverse-phase
Sonnich Meier et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 145(3), 420-430 (2007-03-09)
Offshore oil production releases large amounts of lipophilic compounds in produced water into the ocean. In 2004, 143 million m(3) produced water, containing approximately 13 tons of long-chain (>C(4)) alkylphenols (AP), was discharged from installations in the Norwegian sector of
J Pablo Lamas et al.
Analytical and bioanalytical chemistry, 394(8), 2231-2239 (2009-07-07)
A method for the determination of dimethyl fumarate (DMF), benzothiazole (BT) and tert-butylphenol (TBP) in desiccant and antimould agents employed for protecting consumer products from humidity and mould has been developed. The method is based on ultrasound-assisted extraction (UAE) followed
Pierre Etienne Chabrier et al.
CNS drug reviews, 13(3), 317-332 (2007-09-27)
BN82451 belongs to a new family of small molecules designated as multitargeting or hybrid molecules. BN82451 is orally active, has good central nervous system penetration, and elicits potent neuronal protection and antiinflammatory properties. Neuronal protection is due to Na+ channel
Kai K Lie et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 150(2), 141-149 (2009-04-22)
The release of produced water (PW), a by-product of offshore oil production, has increased in Norwegian waters in recent years. Alkylphenols (AP), a major component of PW, have been shown to have endocrine disrupting effects on several fish species. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service