Skip to Content
Merck
All Photos(1)

Key Documents

W387101

Sigma-Aldrich

4-Methyl-2-oxopentanoic acid sodium salt

97%

Synonym(s):

Sodium 4-methyl-2-oxovalerate, α-Ketoisocaproic acid sodium salt, 4-Methyl-2-oxopentanoic acid sodium salt, 4-Methyl-2-oxovaleric acid sodium salt, Ketoleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCOONa
CAS Number:
Molecular Weight:
152.12
FEMA Number:
3871
Beilstein:
4239297
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Assay

97%

mp

275 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

buttery

SMILES string

[Na+].CC(C)CC(=O)C([O-])=O

InChI

1S/C6H10O3.Na/c1-4(2)3-5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1

InChI key

IXFAZKRLPPMQEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Wax et al.
Journal of dietary supplements, 10(1), 6-16 (2013-01-30)
Glycine-arginine-α-ketoisocaproic acid (GAKIC) has been proposed to increase anaerobic high-intensity exercise performance in male subjects. However, the effects of GAKIC ingestion in female subjects have not been studied. Therefore, the purpose of this study was to investigate the effects of
Li Qi et al.
Talanta, 81(4-5), 1554-1559 (2010-05-06)
A new strategy for the enantioseparation of D,L-amino acids employing the principle of ligand exchange capillary electrophoresis with Zn(II)-L-valine complex as a chiral selecting system in the presence of beta-cyclodextrin has been designed. Successful enantioseparation of label free and labeled
Yuhong Zhu et al.
Enzyme and microbial technology, 49(4), 321-325 (2011-11-25)
This work aims to produce α-ketoisocaproate (KIC) from L-leucine via the free-whole-cell biotransformation of Rhodococcus opacus DSM 43250. The effects of temperature, pH, substrate concentration, cell concentration, and rotating speed on KIC production were examined. Furthermore, the biotransformation conditions were
William S Crim et al.
Molecular and cellular endocrinology, 323(2), 246-255 (2010-03-10)
The antioxidant and anti-inflammatory compound AGI-1067 (succinobucol) has potential as an oral anti-diabetic agent. AGI-1067 reduces H(b)A1c, improves fasting plasma glucose, and reduces new-onset diabetes. We investigated AGI-1067 for possible effects on mouse pancreatic islets in vitro. Pretreatment with 10
Nelo Eidy Zanchi et al.
Amino acids, 40(4), 1015-1025 (2010-07-08)
Amino acids such as leucine and its metabolite α-ketoisocaproate (KIC), are returning to be the focus of studies, mainly because of their anti-catabolic properties, through inhibition of muscle proteolysis and enhancement of protein synthesis. It is clear that these effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service