Skip to Content
Merck
All Photos(1)

Documents

W365009

Sigma-Aldrich

Butyl salicylate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
3650
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.763

biological source

synthetic

Assay

≥99%

form

liquid

refractive index

n20/D 1.5120 (lit.)

bp

268 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

clover

SMILES string

CCCCOC(=O)c1ccccc1O

InChI

1S/C11H14O3/c1-2-3-8-14-11(13)9-6-4-5-7-10(9)12/h4-7,12H,2-3,8H2,1H3

InChI key

YFDUWSBGVPBWKF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Chen et al.
Electrophoresis, 38(22-23), 3036-3047 (2017-07-18)
In the present study, a monolithic capillary column with higher permeability was developed for the in vivo discrimination of four coumarin analogs (bergapten, 2'-acetylangelicin, imperatorin, and osthole) that typically require long separation times in HPLC. Instead of conventional methacrylate ester
Wei-Chan Cui et al.
Insect biochemistry and molecular biology, 97, 31-39 (2018-04-27)
Plant volatiles are vital cues in the location of hosts for feeding and oviposition for Lepidoptera moths. The noctuid Helicoverpa assulta is a typical polyphagous moth, regarded as a good model for studying the olfactory reception of plant volatiles. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service