Skip to Content
Merck
All Photos(1)

Key Documents

W329509

Sigma-Aldrich

DL-Isoleucine

99%, FCC, FG

Synonym(s):

DL-2-Amino-3-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
FEMA Number:
3295
Beilstein:
1721790
EC Number:
Council of Europe no.:
10127
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.010
NACRES:
NA.21
grade:
FG
biological source:
synthetic
Pricing and availability is not currently available.

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC

Assay

99%

mp

290 °C (dec.) (lit.)

application(s)

flavors and fragrances

Organoleptic

odorless

SMILES string

CC[C@@H](C)[C@@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m1/s1

InChI key

AGPKZVBTJJNPAG-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A mixture of 4 stereoisomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service